To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:lin-28 homolog B (C. elegans)
- Antigen Symbol:LIN28B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western Blot:Yes
- Size:200 ul
- Environmentally Preferable:
- Gene ID:Q6ZN17
- Antigen Synonyms:LIN28B
- Storage Buffer:PBS, 0.09% sodium azide
- Storage Temperature:Antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles.
- Concentration:0.25 mg/mL
- Shipping Temperature:4 °C
- Immunogen:KLH-conjugated synthetic peptide between amino acids 1-30 from N-terminal region of human LIN28B.
- Tested Applications:Fluorescence-Activated Cell Sorting
- Purification:Protein A purified
- Cat. No.:10074-112
- Supplier no.:70-948
LIN28B Antibody can be used for ELISA starting at 1:1000, Flow Cytometry starting at 1:10, Immunohistochemistry in paraffin embeded tissues starting at 10?g/mL, and Western Blotting starting at 1:50 - 1:100.
PBS, 0.09% sodium azide
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: LIN28B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human