Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MUC1 Mouse Monoclonal Antibody [clone: [GP1.4]]
Anti-MUC1 Mouse Monoclonal Antibody [clone: [GP1.4]]
Catalog # 10073-656
Supplier:  Prosci
undefined
Anti-MUC1 Mouse Monoclonal Antibody [clone: [GP1.4]]
Catalog # 10073-656
Supplier:  Prosci
Supplier Number:  70-048

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    mucin 1, cell surface associated
  • Antigen Symbol:
    MUC1
  • Clonality:
    Monoclonal
  • Clone:
    [GP1.4]
  • Conjugation:
    Unconjugated
  • Flow Cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Form:
    Liquid
  • Gene ID:
    P15941
  • Antigen Synonyms:
    MUC1
  • Storage Buffer:
    PBS containing 0.09% sodium azide
  • Storage Temperature:
    MUC1 Monoclonal antibody may be stored short term at 4 °C, long term aliquot and store at –20 °C, avoid freeze thaw cycles. Store undiluted.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Human milk fat globule
  • Tested Applications:
    Immunocytochemistry
  • Purification:
    Purified
  • Cat. No.:
    10073-656
  • Supplier no.:
    70-048

Specifications

About this item

MUC1 Monoclonal Antibody can be used for flow cytometry starting at 1:25 - 1:200, immunohistochemistry (tested on frozen tissues starting at 1:25 - 1:200, immunohistochemistry (tested on parafin embedded tissues) starting at 10 ?g/mL and Western Blot starting at 1:100 - 1:1000.

PBS containing 0.09% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: [GP1.4]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human