Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-MAP2K5 Rabbit Polyclonal Antibody
Anti-MAP2K5 Rabbit Polyclonal Antibody
Catalog # 10075-500
Supplier:  Prosci
undefined
Anti-MAP2K5 Rabbit Polyclonal Antibody
Catalog # 10075-500
Supplier:  Prosci
Supplier Number:  50-245

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    mitogen activated protein kinase kinase 5
  • Antigen Symbol:
    Map2k5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Zebrafish,
    Xenopus,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Form:
    Liquid
  • Gene ID:
    Q62862
  • Antigen Synonyms:
    Map2k5
  • Modification:
    Ser 311/Thr 315
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    49
  • Storage Temperature:
    MEK5 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser311, Thr315 of MEK5.
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-500
  • Supplier no.:
    50-245

Specifications

About this item

MEK5 (also known as MKK5) is a dual specificity serine/threonine protein kinase belonging to the MAP kinase kinase family. MEK5 has been shown to specifically activate ERK5 (Zhou et al., 1995) whereas MEK5 itself is regulated by MEKK3 (Chao et al., 1999). An important link between MEK5 and metastatic prostrate cancer has been demonstrated (Mehta et.al., 2003). Dual phosphorylation of Ser311 and Thr315 have been implicated in cell proliferation (Cameron et al., 2004).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with mouse, human, chicken, bovine, canine, non-human primate, Xenopus, and Zebrafish tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Map2k5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Bovine, Chcicken, Xenopus