Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-RAF1 Rabbit Polyclonal Antibody
Catalog # 10075-444
Supplier:  Prosci
undefined
Anti-RAF1 Rabbit Polyclonal Antibody
Catalog # 10075-444
Supplier:  Prosci
Supplier Number:  50-238

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    v-raf-leukemia viral oncogene 1
  • Antigen Symbol:
    Raf1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Form:
    Liquid
  • Gene ID:
    P11345
  • Antigen Synonyms:
    Raf1
  • Modification:
    Ser 642
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    74
  • Storage Temperature:
    Raf-1 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser642 of rat Raf-1.
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-444
  • Supplier no.:
    50-238

Specifications

About this item

The Ras pathway is a critical signal transduction cascade involved in regulating cellular proliferation, differentiation, survival, and oncogenic transformation. Members of the Raf serine/threonine kinase family are key intermediates in this cascade, functioning to relay signals from activated Ras to the downstream protein kinases MEK and ERK (Marshall, 1996). Previous studies have shown that phosphorylation is required for Raf-1 activation (Dhillon and Kolch, 2002; Chong et al., 2003). Recent work has demonstrated that phosphorylation also regulates the downregulation of Raf (Dougherty et al., 2005) with two sites participating: Ser301 and Ser642.

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, canine, chicken, human, mouse, and non human primates based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Raf1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine, Chicken