Specifications
- Antibody Type:Primary
- Antigen Name:glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen Symbol:Grin2b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q00960
- Antigen Synonyms:Grin2b
- Modification:Tyr 1336
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:180
- Storage Temperature:NMDA antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:Phosphopeptide corresponding to amino acid residues surrounding the phospho-Tyr1336 of the NR2B subunit of the rat NMDA receptor.
- Purification:Affinity Purified
- Cat. No.:10075-738
- Supplier no.:50-233
Specifications
About this item
The NMDAR plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death (Grosshans et al., 2002; Wenthold et al., 2003; Carroll and Zukin, 2002). The rat NMDAR1 (NR1) was the first subunit of the NMDAR to be cloned. The NR1 protein can form NMDA activated channels when expressed in Xenopus oocytes but the currents in such channels are much smaller than those seen in situ. Channels with more physiological characteristics are produced when the NR1 subunit is combined with one or more of the NMDAR2 (NR2 A-D) subunits (Ishii et al., 1993). Phosphorylation of Tyr1336 is thought to potentiate NMDA receptor-dependent influx of calcium (Takasu et al., 2002) and ischemia may also increase the phosphorylation of this site (Takagi et al., 2003).
The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with human, mouse and non-human primate tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Grin2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat