Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-STXBP2 Rabbit Polyclonal Antibody
Anti-STXBP2 Rabbit Polyclonal Antibody
Catalog # 10075-734
Supplier:  Prosci
undefined
Anti-STXBP2 Rabbit Polyclonal Antibody
Catalog # 10075-734
Supplier:  Prosci
Supplier Number:  50-230

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    syntaxin binding protein 2
  • Antigen Symbol:
    Stxbp2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Xenopus,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Form:
    Liquid
  • Gene ID:
    Q62753
  • Antigen Synonyms:
    Stxbp2
  • Modification:
    Ser 515
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    65
  • Storage Temperature:
    Munc-18 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser515 of Munc-18.
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-734
  • Supplier no.:
    50-230

Specifications

About this item

Munc-18 ( mammalian homologue of Unc-18) is a protein that is thought to be involved in regulating exocytosis due, at least in part, to it ability to bind syntaxin (Ciufo et al., 2005). Munc18-1 is a neuron-specific member of the Sec1/Munc18 protein family that binds to syntaxin1A and is thought to stabilize the complex (Liu et al., 2004). The function of Munc-18 is thought to be regulated by PKC phosphorylation of Ser515 on the Munc-18 protein (Sassa et al., 1996).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with bovine, canine, chicken, human, mouse, non-human primates and Xenopus based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Stxbp2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine, Chicken, Xenopus