Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GSK3B Rabbit Polyclonal Antibody
Anti-GSK3B Rabbit Polyclonal Antibody
Catalog # 10075-626
Supplier:  Prosci
undefined
Anti-GSK3B Rabbit Polyclonal Antibody
Catalog # 10075-626
Supplier:  Prosci
Supplier Number:  50-228

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    glycogen synthase kinase 3 beta
  • Antigen Symbol:
    Gsk3b
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Zebrafish,
    Xenopus,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Form:
    Liquid
  • Gene ID:
    P18266
  • Antigen Synonyms:
    Gsk3b
  • Modification:
    Ser 9
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    46
  • Storage Temperature:
    Glycogen synthase antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser9 of GSK3b.
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-626
  • Supplier no.:
    50-228

Specifications

About this item

Glycogen synthase kinase 3 (GSK3) is a serine/threonine kinase that is involved in the regulation of many signaling pathways. To date, 2 isoforms have been identified: GSK3alpha and GSK3beta. Specifically, GSK3beta has been shown to play a key inhibitory role in both the insulin and Wnt signaling pathways (Papkoff and Aikawa 1998). It has been suggested that Ser9 phosphorylation underlies the inhibition of GSK3beta by insulin (Sutherland et al., 1993).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will react with human, mouse, chicken, bovine, canine, non-human primate, Zebra fish and Xenopus based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Gsk3b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Bovine, Chcicken, Xenopus