Specifications
- Antibody Type:Primary
- Antigen Name:solute carrier family 6 (neurotransmitter transporter), member 3
- Antigen Symbol:SLC6A3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q01959
- Antigen Synonyms:SLC6A3
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:88
- Storage Temperature:Dopamine Transporter antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:Peptide from the intracellular C-terminus region of human dopamine transporter (DAT), conjugated to keyhole limpet hemocyanin (KLH).
- Purification:Affinity Purified
- Cat. No.:10075-344
- Supplier no.:50-204
Specifications
About this item
The dopamine transporter (DAT) is responsible for the reaccumulation of dopamine after it has been released. DAT antibodies and antibodies for other markers of catecholamine biosynthesis are widely used as markers for dopaminergic and noradrenergic neurons in a variety of applications including depression, schizophrenia, Parkinson’s disease and drug abuse (Kish et al., 2001; Zhu et al., 2000; Zhu et al., 1999). Levels of DAT protein expression are altered by chronic drug administration (Wilson et al., 1996).
The antibody has been tested in Western blots of SDS-solubilized human and mouse striatal samples and in IHC applications with monkey (Macaque) brain sections.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SLC6A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse