Specifications
- Antibody Type:Primary
- Antigen Name:gap junction protein, alpha 1
- Antigen Symbol:Gja1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Xenopus,Sheep,Chicken,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P08050
- Antigen Synonyms:Gja1
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:43
- Storage Temperature:Connexin antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:Peptide corresponding to amino acid residues from the C-terminal region of rat connexin43.
- Purification:Affinity Purified
- Cat. No.:10075-466
- Supplier no.:50-200
Specifications
About this item
Gap junctional intercellular communication is thought to play a key role in development and may also be involved in epilepsy (Aronica et al., 2001). Connexin43 forms gap-junctional channels and regulates the permeability of these gap junctions to small organic molecules. Permeability of connexin43 is known to be regulated by phosphorylation at er368 by protein kinase C (Yogo et al., 2002; Bao et al., 2004a). Phosphorylation of Ser368 by PKC induces a conformational change of connexin43 that results in a decrease in gap junction permeability (Bao et al., 2004b).
The antibody has been directly tested for reactivity in Western blots in rat tissues. It is anticipated that the antibody will also work with chicken, dog, finch, human, mouse, non-human primate, sheep, Xenopus and zebra fish tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Gja1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Sheep, Chicken, Xenopus