Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-NSF Rabbit Polyclonal Antibody
  10075-476
 :  Prosci
undefined
Anti-NSF Rabbit Polyclonal Antibody
  10075-476
 :  Prosci
 :  50-183

 

  • Antibody Type:
    Primary
  • Antigen Name:
    N-ethylmaleimide-sensitive factor
  • Antigen Symbol:
    Nsf
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Form:
    Liquid
  • Gene ID:
    Q9QUL6
  • Antigen Synonyms:
    Nsf
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    75
  • Storage Temperature:
    NSF antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Peptide corresponding to amino acid residues from the C-terminal region of rat NSF, conjugated to keyhole limpet hemocyanin (KLH).
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-476
  • Supplier no.:
    50-183

 

 

NSF (N-ethylmaleimide sensitive fusion protein) is a critical component of the SNARE (soluble NSF attachment protein receptors) protein complex that is involved in synaptic vesicle trafficking. Specifically, NSF has been found to be essential in membrane fusion. Furthermore, NSF has been recently demonstrated to bind other protein complexes such as AMPA receptor subunits (GluR2), GATE-16, LMA-1 and Rabs suggesting a more diverse role in the assembly of various protein complexes (Whiteheart et al., 2004).

The antibody has been directly tested for reactivity in rat tissue. It is anticipated that the antibody will also work with human and mouse based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Nsf
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat