- Antibody Type:Primary
- Antigen Name:discs, large homolog 4 (Drosophila)
- Antigen Symbol:Dlg4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Zebrafish,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P31016
- Antigen Synonyms:Dlg4
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:95
- Storage Temperature:PSD-95 antibody can be stored at –20 °C and is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:Peptide corresponding to amino acid residues from the N-terminal region of rat PSD-95, conjugated to keyhole limpet hemocyanin (KLH).
- Purification:Affinity Purified
- Cat. No.:10075-468
- Supplier no.:50-169
PSD-95 is a very prominent component of the postsynaptic densities of synapses. It contains three PDZ domains which play key roles in its interactions with other proteins in the synapse. It has been proposed that these PDZ domains organize glutamate receptors and their associated signaling proteins and determine the size and strength of synapses (Kim and Sheng, 2004). Recent work suggests that interaction of the NMDAR with PSD-95 via these PDZ domains can be regulated by phosphorylation (Chung et al., 2004).
The antibody has been directly tested for reactivity in Western blots in rat and mouse tissues. It is anticipated that the antibody will also work with bovine, human, mouse, non-human primates and zebra fish tissues based on the fact that these species have 100% homology with the amino acid sequence used as antigen.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Dlg4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Bovine, Zebrafish