Anti-RHO Mouse Polyclonal Antibody [clone: 1D4]
Catalog # 10075-566
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:rhodopsin
- Antigen Symbol:RHO
- Clonality:Monoclonal
- Clone:1D4
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Amphibian,Mammals
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P02699
- Antigen Synonyms:RHO
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:39
- Shipping Temperature:4 °C
- Immunogen:Bovine rhodopsin.
- Purification:Protein G purified
- Cat. No.:10075-566
- Supplier no.:50-141
Specifications
About this item
Protein G purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: RHO
Clonality: Monoclonal
Clone: 1D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mammal, Amphibian