Anti-GRIN2B Rabbit Polyclonal Antibody
10075-570
: Prosci
- Antibody Type:Primary
- Antigen Name:glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen Symbol:Grin2b
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q00960
- Antigen Synonyms:Grin2b
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:180
- Shipping Temperature:4 °C
- Immunogen:Peptide from the N-terminus of the NR2B subunit of rat NMDA receptor.
- Purification:Affinity Purified
- Cat. No.:10075-570
- Supplier no.:50-131
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: Grin2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat