Order Entry
United States
ContactUsLinkComponent
Anti-GRIN2C Rabbit Polyclonal Antibody
Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # 10075-636
Supplier:  Prosci
undefined
Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # 10075-636
Supplier:  Prosci
Supplier Number:  50-114
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    glutamate receptor, ionotropic, N-methyl D-aspartate 2C
  • Antigen Symbol:
    Grin2c
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    0.1 mL
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q00961
  • Antigen Synonyms:
    Grin2c
  • Modification:
    Ser 1244
  • Storage Buffer:
    100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
  • Molecular Weight:
    140
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Synthetic phospho-peptide surrounding the phospho Ser1244 of the NR2C-subunit of rat NMDA receptor.
  • Purification:
    Affinity Purified
  • Cat. No.:
    10075-636
  • Supplier no.:
    50-114

Specifications

About this item

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type: Primary
Antigen: Grin2c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat