Anti-GRIN2C Rabbit Polyclonal Antibody
Catalog # 10075-634
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:glutamate receptor, ionotropic, N-methyl D-aspartate 2C
- Antigen Symbol:Grin2c
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:Q00961
- Antigen Synonyms:Grin2c
- Modification:Ser 1096
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:140
- Shipping Temperature:4 °C
- Immunogen:Synthetic phospho-peptide surrounding the phospho Ser1096 of the NR2C-subunit of rat NMDA receptor.
- Purification:Affinity Purified
- Cat. No.:10075-634
- Supplier no.:50-113
Specifications
About this item
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Type: Primary
Antigen: Grin2c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat