Specifications
- Antibody Type:Primary
- Antigen Name:glutamate receptor, metabotropic 1
- Antigen Symbol:Grm1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Rat
- Western Blot:Yes
- Size:0.1 mL
- Form:Liquid
- Gene ID:P23385
- Antigen Synonyms:Grm1
- Storage Buffer:100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
- Molecular Weight:125/250
- Storage Temperature:Metabotropic Glutamate Receptor 1a antibody can be stored at –20 °C. After reconstitution in 50 μl PBS, the antibody should be aliquot and stored at –20 °C. This product is stable at –20 °C for at least 1 year.
- Shipping Temperature:4 °C
- Immunogen:mGluR 1a polyclonal antibody was raised against a peptide from the C-terminal region of mGluR1a.
- Purification:Affinity Purified
- Cat. No.:10075-606
- Supplier no.:50-101
Specifications
About this item
The metabotropic glutamate receptors (mGluRs) are key receptors in the modulation of excitatory synaptic transmission in the central nervous system. They are implicated in many forms of neural plasticity as well as learning and memory and drug abuse. Group I metabotropic glutamate receptors (consisting of mGluR1 and mGluR5) are G-protein-coupled neurotransmitter receptors that are localized in the perisynaptic region of the postsynaptic membrane. When activated, Group I mGluRs lead to stimulation of phospholipase and activation of Protein Kinase C. In contrast activation of Group II metabotropic receptors (mGluR2 and mGluR3) leads to inhibition of adenylate cyclase.
Metabotropic Glutamate Receptor 1a antibody allows for strong and specific immunolabeling of ~125k mGluR1a and the ~250k mGluR1a dimer in Western blots of rat brain at antibody dilution of 1:1000. Immunolabeling blocked by preadsorption of antibody with the C-terminal peptide immunogen. Applications include Dot Blots (DB), Immunohistochemistry (IHC) and Western Blots (WB). Rabbit anti-mGluR recognizes mGluR1a in rat brain. When internally tested under ideal conditions the working dilutions were 1:500 for IHC and 1:1000 for DB and WB.
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Reconstitute in 50 ?l PBS. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Grm1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat