Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-RAMP3 Rabbit Polyclonal Antibody
Anti-RAMP3 Rabbit Polyclonal Antibody
Catalog # 10078-430
Supplier:  Prosci
undefined
Anti-RAMP3 Rabbit Polyclonal Antibody
Catalog # 10078-430
Supplier:  Prosci
Supplier Number:  48-972

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    receptor (G protein-coupled) activity modifying protein 3
  • Antigen Symbol:
    RAMP3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.05 mg
  • Form:
    Liquid
  • Gene ID:
    O60896
  • Antigen Synonyms:
    RAMP3
  • Storage Buffer:
    PBS, 0.01% sodium azide, 50% glycerol.
  • Storage Temperature:
    Short term: 4 °C. Long term: Store at –20 °C. Avoid freeze-thaw cycles.
  • Concentration:
    0.5 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    RAMP3 antibody was raised against a synthetic peptide corresponding to N-Terminal residues of RAMP3 (Human).
  • Tested Applications:
    Immunoaffinity Chromatography
  • Purification:
    Immunoaffinity Chromatography
  • Cat. No.:
    10078-430
  • Supplier no.:
    48-972

Specifications

About this item

RAMP3 is a member of the RAMP family of single-transmembrane-domain proteins, called receptor-activity-modifying proteins. RAMPs are type I transmembrane proteins with an extracellular N terminus and a cytoplasmic C terminus. RAMPs are required to transport calcitonin-receptor-like receptor (CRLR) to the plasma membrane. CRLR, a receptor with seven transmembrane domains, can function as either a calcitonin-gene-related peptide (CGRP) receptor or an adrenomedullin receptor, depending on which members of the RAMP family are expressed. Co-expression of RAMP3 with CRLR results in the production of a functional adrenomedullin receptor.

RAMP3 antibody can be used in ELISA starting at 1:3000 - 1:6000, Western Blot starting at 1:3000 - 1:6000, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation

PBS, 0.01% sodium azide, 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RAMP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human