Specifications
- Antibody Type:Primary
- Antigen Name:NIMA-related kinase 2
- Antigen Symbol:NEK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:0.05 mg
- Form:Liquid
- Gene ID:P51955
- Antigen Synonyms:NEK2
- Storage Buffer:0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, 0.01% sodium azide.
- Storage Temperature:Store NEK2 antibody at –20 °C prior to opening. Dilute only prior to immediate use. For extended storage aliquot contents and freeze at –20 °C or below. As with all antibodies avoid freeze/thaw cycles.
- Concentration:1 mg/mL
- Shipping Temperature:4 °C
- Immunogen:NEK2 antibody was raised against amino acids 287 - 299 of NEK2 (Human).
- Tested Applications:Immunoaffinity Chromatography
- Purification:Immunoaffinity Chromatography
- Cat. No.:10078-252
- Supplier no.:48-615
Specifications
About this item
NEK2, also called serine/threonine-protein kinase Nek2NimA-related protein kinase 2, NimA-like protein kinase 1 and HSPK 21, is a protein kinase involved in mitotic regulation that may have a role at the G2-M transition and may also play a role in meiosis. NEK2 has a nuclear localization and accumulates throughout S phase and shows maximal levels in late G2. This expression pattern is highly reminiscent of that of A and B cyclins.
NEK2 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.
0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.
Store NEK2 antibody at -20 °C prior to opening. Dilute only prior to immediate use. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: NEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human