Anti-GALNS Goat Polyclonal Antibody
Catalog # 10116-998
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:galactosamine (N-acetyl)-6-sulfate sulfatase
- Antigen Symbol:GALNS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:P34059
- Antigen Synonyms:GALNS
- Storage Buffer:GALNS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:GALNS antibody was raised against a 12 amino acid synthetic peptide near the internal region of GALNS.
- Purification:GALNS antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10116-998
- Supplier no.:46-663
Specifications
About this item
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 60 kDa band observed in human bone marrow and placenta lysates (calculated MW of 58.0 kDa according to NP_000503.1). Recommended concentration: 0.01-0.03 ?g/mL.
GALNS antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GALNS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human