Specifications
- Antibody Type:Primary
- Antigen Name:Rac GTPase activating protein 1
- Antigen Symbol:RACGAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q9H0H5
- Antigen Synonyms:RACGAP1
- Storage Buffer:RACGAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:RACGAP1 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of RACGAP1.
- Purification:RACGAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10116-560
- Supplier no.:46-260
Specifications
About this item
ELISA: Antibody detection limit dilution 1:32000. Western Blot: Approximately 60 kDa band observed in human testis lysates (calculated MW of 71.0 kDa according to NP_037409.2). The observed molecular weight corresponds to earlier findings in literature with different antibodies (Toure et al., J Biol Chem 1998 Mar 13;273 (11) :6019-23; PMID: 9497316). Recommended concentration: 0.1-0.3 ?g/mL. Approximately 75 kDa band observed in K562 and Jurkat lysates (calculated MW of 71 kDa according to NP_037409). Recommended concentration: 1-3 ?g/mL. Immunohistochemistry: In paraffin embedded human testis, shows nuclear staining of many cells. Recommended concentration, 1 ?g/mL.
RACGAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RACGAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human