Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-PRAM1 Goat Polyclonal Antibody
Anti-PRAM1 Goat Polyclonal Antibody
Catalog # 10116-500
Supplier:  Prosci
undefined
Anti-PRAM1 Goat Polyclonal Antibody
Catalog # 10116-500
Supplier:  Prosci
Supplier Number:  46-214

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    PML-RARA regulated adaptor molecule 1
  • Antigen Symbol:
    PRAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    Q96QH2
  • Antigen Synonyms:
    PRAM1
  • Storage Buffer:
    PRAM1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    PRAM1 antibody was raised against a 15 amino acid synthetic peptide near the C-Terminus of PRAM1.
  • Purification:
    PRAM1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10116-500
  • Supplier no.:
    46-214

Specifications

About this item

ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 90-100 kDa band seen in Jurkat cell lysates. Recommended for use at 1-3 ?g/mL. Overnight incubation is recommended with this antibody. Please note that although the predicted MW of PRAM-1 is 79 kDa, Moog-Lutz et al., (J. Biol. Chem, Jun 2001; 276: 22375 - 22381) detected aPRAM-1 migrating with an apparent molecular mass of 97 kDa and this is what we observed as well.

PRAM1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PRAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human