Anti-Hap1 Goat Polyclonal Antibody
10113-322
: Prosci
- Antibody Type:Primary
- Antigen Name:huntingtin-associated protein 1
- Antigen Symbol:Hap1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:O35668
- Antigen Synonyms:Hap1
- Storage Buffer:HAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:HAP1 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of HAP1.
- Purification:HAP1 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10113-322
- Supplier no.:45-722
ELISA: Antibody detection limit dilution 1:32,000. Western Blot: Approximately 110 kDa band in mouse and rat brain lysates after 0.5 ?g/mL antibody staining.8 kDa according to NP_034534.1. The 110 kDa band was successfully blocked by incubation with the immunizing peptide.
HAP1 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Hap1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat