Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GRIN3B Goat Polyclonal Antibody
Anti-GRIN3B Goat Polyclonal Antibody
Catalog # 10114-700
Supplier:  Prosci
undefined
Anti-GRIN3B Goat Polyclonal Antibody
Catalog # 10114-700
Supplier:  Prosci
Supplier Number:  42-626

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    glutamate receptor, ionotropic, N-methyl-D-aspartate 3B
  • Antigen Symbol:
    GRIN3B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    O60391
  • Antigen Synonyms:
    GRIN3B
  • Storage Buffer:
    GRIN3B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
  • Storage Temperature:
    Aliquot and store at –20 °C. Minimize freezing and thawing.
  • Concentration:
    500 ug/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    GRIN3B antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of GRIN3B.
  • Purification:
    GRIN3B antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Cat. No.:
    10114-700
  • Supplier no.:
    42-626

Specifications

About this item

ELISA: Antibody detection limit dilution 1:8000. Western Blot: Approximately 110 kDa band observed in human brain (cerebellum and cerebral cortex) lysates (calculated MW of 113 kDa according to NP_619635.1). Recommended concentration: 0.5-1.5 ?g/mL.

GRIN3B antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GRIN3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human