Anti-SFRP4 Goat Polyclonal Antibody
Catalog # 10113-986
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:secreted frizzled-related protein 4
- Antigen Symbol:SFRP4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:Q6FHJ7
- Antigen Synonyms:SFRP4
- Storage Buffer:SFRP4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% bovine serum albumin.
- Storage Temperature:Aliquot and store at –20 °C. Minimize freezing and thawing.
- Concentration:500 ug/mL
- Shipping Temperature:4 °C
- Immunogen:SFRP4 antibody was raised against a 15 amino acid synthetic peptide near the internal region of SFRP4.
- Purification:SFRP4 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:10113-986
- Supplier no.:42-066
Specifications
About this item
ELISA: Antibody detection limit dilution 1:16000. Western Blot: Approximately 40 kDa band observed in human uterus lysates (calculated MW of 39.8 kDa according to NP_003005.2). Recommended concentration: 0.3-1 ?g/mL. Immunohistochemistry: In paraffin embedded human skeletal muscle shows interfibrillar staining. Recommended concentration, 4-6 ?g/mL.
SFRP4 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SFRP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human