Anti-Clostridum difficile Mouse Monoclonal Antibody [clone: 5155]
Catalog # 10070-800
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Clostridum difficile toxin Antibody [5155]
- Antigen Symbol:Clostridum difficile
- Clonality:Monoclonal
- Clone:5155
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Bacteria
- Size:0.1 mg
- Form:Liquid
- Storage Buffer:Clostridum difficile toxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
- Storage Temperature:Clostridum difficile toxin antibody can be stored at –20 °C, stable for one year.
- Shipping Temperature:4 °C
- Purification:Clostridum difficile toxin antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.
- Cat. No.:10070-800
- Supplier no.:35-598
Specifications
About this item
Clostridum difficile toxin antibody is purified from ascites fluid or culture medium by protein A chromoatography or sequential differential precipitations.
Clostridum difficile toxin antibody is in a phosphate saline buffer (0.01M, pH 7.2) containing 0.1% sodium azide preservative. No stabilizing proteins have been added.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Clostridum difficile toxin Antibody [5155]
Clonality: Monoclonal
Clone: 5155
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Bacteria