Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-DSTYK Rabbit Polyclonal Antibody
  10062-546
 :  Prosci
undefined
Anti-DSTYK Rabbit Polyclonal Antibody
  10062-546
 :  Prosci
 :  3291

 

  • Antibody Type:
    Primary
  • Antigen Name:
    receptor (TNFRSF)-interacting serine-threonine kinase 1
  • Antigen Symbol:
    RIPK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    0.1 mg
  • Form:
    Liquid
  • Gene ID:
    Q13546
  • Antigen Synonyms:
    RIPK1
  • Storage Buffer:
    RIPK1 Antibody is supplied in PBS containing 0.02% sodium azide.
  • Molecular Weight:
    Predicted: 74 kDa
  • Storage Temperature:
    RIPK1 antibody can be stored at 4 °C for three months and –20 °C, stable for up to one year.
  • Concentration:
    1 mg/mL
  • Shipping Temperature:
    4 °C
  • Immunogen:
    RIPK1 antibody was raised against a 14 amino acid peptide near the carboxy terminus of human RIP1. The immunogen is located within amino acids 600 - 650 of RIPK1.
  • Purification:
    RIPK1 Antibody is affinity chromatography purified via peptide column.
  • Cat. No.:
    10062-546
  • Supplier no.:
    3291

 

 

RIP1 Antibody: RIP1 (Receptor Interacting Protein), also known as RIPK1, is a crucial 74 kD adaptor kinase in several of stress-induced signaling pathways and on the crossroad of a cell’s decision to live or die. RIP1 contains an N-terminal region with homology to protein kinases, an intermediate domain capable of association with MAPKKK and a C-terminal region containing a death domain motif present in the Fas and TNFR1 intracellular domains. Full length RIP1 is important for signallling to NF-kappa-B, MAPKs and necrosis, whereas caspase-8 generates a C-terminal RIP1 cleavage fragment, promoting TNF-induced apoptosis. It is required for TNFRSF1A-mediated and TLR3-induced NF-kappa-B activation. RIP1-deficient mice fail to thrive, displaying extensive apoptosis in both lymphoid and adipose tissues and dying at 1-3 days of age.

RIP1 antibody can be used for detection of RIP1 by immunohistochemistry at 5 - 10 ?g/mL.

RIP1 Antibody is affinity chromatography purified via peptide column.

RIP1 Antibody is supplied in PBS containing 0.02% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RIPK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human