- Antibody Type:Primary
- Antigen Name:GA binding protein transcription factor, alpha subunit 60kDa
- Antigen Symbol:GABPA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q06546
- Antigen Synonyms:GABPA
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:51 kDa, 97 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store GABPA antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GABPA.
- Purification:Antibody is supplied as total IgG.
- Cat. No.:10111-336
- Supplier no.:31-197
GA Binding Protein. chain (GABP-. subunit, GABPA, nuclear respiratory factor-2 subunit. transcription factor E4TF1-60) is one of three GA-binding protein transcription factor subunits which functions as a DNA-binding subunit. Since this subunit shares identity with a subunit encoding the nuclear respiratory factor 2 gene, it is likely involved in activation of cytochrome oxidase expression and nuclear control of mitochondrial function. This subunit also shares identity with a subunit constituting the transcription factor E4TF1, responsible for expression of the adenovirus E4 gene. Because of its chromosomal localization and ability to form heterodimers with other polypeptides, this gene may play a role in the Down Syndrome phenotype.
GABPA antibody can be used for detection of GABPA by ELISA at 1:312500. GABPA antibody can be used for detection of GABPA by western blot at 0.5-1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is supplied as total IgG.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 200 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GABPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish