Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-GK2 Rabbit Polyclonal Antibody
Anti-GK2 Rabbit Polyclonal Antibody
Catalog # 10110-950
Supplier:  Prosci
undefined
Anti-GK2 Rabbit Polyclonal Antibody
Catalog # 10110-950
Supplier:  Prosci
Supplier Number:  30-940

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    glycerol kinase 2
  • Antigen Symbol:
    GK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 ul
  • Form:
    Liquid
  • Gene ID:
    Q01415
  • Antigen Synonyms:
    GK2
  • Storage Buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • Molecular Weight:
    61 kDa
  • Storage Temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store GK2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GK2.
  • Purification:
    Antibody is purified by peptide affinity chromatography method.
  • Cat. No.:
    10110-950
  • Supplier no.:
    30-940

Specifications

About this item

GK2 belongs to the FGGY kinase family. GK2 is a key enzyme in the regulation of glycerol uptake and metabolism.

GK2 antibody can be used for detection of GK2 by ELISA at 1:1562500. GK2 antibody can be used for detection of GK2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human