- Antibody Type:Primary
- Antigen Name:caudal
- Antigen Symbol:cad
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Drosophila
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P09085
- Antigen Synonyms:CAD
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:46 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store CAD antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of Drosophila CAD.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10109-928
- Supplier no.:30-324
Cad regulates embryonic abdominal segment formation by zygotically activating expression of knirps (kni) and giant (gt). It plays a role in the establishment of the hindgut and in the invagination of the hindgut primordium during gastrulation. These effects on the gut are achieved by acting combinatorially at the posterior of the embryo to activate transcription of different targets, including folded gastrulation (fog), fork head (fkh) and wingless (wg).
CAD antibody can be used for detection of CAD by ELISA at 1:12500. CAD antibody can be used for detection of CAD by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: cad
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Drosophila