Specifications
- Antibody Type:Primary
- Antigen Name:uronyl-2-sulfotransferase
- Antigen Symbol:UST
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9Y2C2
- Antigen Synonyms:UST
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:48 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store UST antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human UST.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10109-718
- Supplier no.:30-214
Specifications
About this item
UST catalyzes the transfer of sulfate to the position 2 of uronyl residues. UST has mainly activity toward iduronyl residues in dermatan sulfate, and weaker activity toward glucuronyl residues of chondroitin sulfate. It has no activity toward desulfated N-resulfated heparin.
UST antibody can be used for detection of UST by ELISA at 1:312500. UST antibody can be used for detection of UST by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: UST
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog