Specifications
- Antibody Type:Primary
- Antigen Name:apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3B
- Antigen Symbol:APOBEC3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9UH17
- Antigen Synonyms:APOBEC3B
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:46 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store APOBEC3B antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human APOBEC3B.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10109-574
- Supplier no.:30-160
Specifications
About this item
APOBEC3B is a member of the cytidine deaminase family. It is thought that these proteins may be RNA editing enzymes and have roles in growth or cell cycle control.This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
APOBEC3B antibody can be used for detection of APOBEC3B by ELISA at 1:62500. APOBEC3B antibody can be used for detection of APOBEC3B by western blot at 0.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: APOBEC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human