Specifications
- Antibody Type:Primary
- Antigen Name:ring finger protein 39
- Antigen Symbol:RNF39
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:A2BEK3
- Antigen Synonyms:RNF39
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:39 kDa, 46 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store RNF39 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human RNF39.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10109-088
- Supplier no.:29-856
Specifications
About this item
Its gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that RNF39 plays a role in an early phase of synaptic plasticity. Its gene lies within the major histocompatibility complex class I region on chromosome 6.This gene lies within the major histocompatibility complex class I region on chromosome 6. Studies of a similar rat protein suggest that this gene encodes a protein that plays a role in an early phase of synaptic plasticity. Alternative splicing results in three transcript variants encoding different isoforms.
RNF39 antibody can be used for detection of RNF39 by ELISA at 1:312500. RNF39 antibody can be used for detection of RNF39 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RNF39
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human