Specifications
- Antibody Type:Primary
- Antigen Name:naked cuticle homolog 1 (Drosophila)
- Antigen Symbol:NKD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q969G9
- Antigen Synonyms:NKD1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:52 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store NKD1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human NKD1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10108-510
- Supplier no.:29-547
Specifications
About this item
In the mouse, Nkd is a Dishevelled -binding protein that functions as a negative regulator of the Wnt-beta-catenin -Tcf signaling pathway.In the mouse, Nkd is a Dishevelled (see DVL1; MIM 601365)-binding protein that functions as a negative regulator of the Wnt (see WNT1; MIM 164820)-beta-catenin (see MIM 116806)-Tcf (see MIM 602272) signaling pathway.
NKD1 antibody can be used for detection of NKD1 by ELISA at 1:62500. NKD1 antibody can be used for detection of NKD1 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: NKD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human