Anti-DCP2 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
DCP2 is necessary for the degradation of mRNAs, both in normal mRNA turnover and in nonsense-mediated mRNA decay. DCP2 removes the 7-methyl guanine cap structure from mRNA molecules, yielding a 5'-phosphorylated mRNA fragment and 7m-GDP. It has higher activity towards mRNAs that lack a poly (A) tail, but has no activity towards a cap structure lacking a RNA moiety.DCP2 is a key component of an mRNA-decapping complex required for removal of the 5-prime cap from mRNA prior to its degradation from the 5-prime end (Fenger-Gron et al., 2005 [PubMed 16364915]).
DCP2 antibody can be used for detection of DCP2 by ELISA at 1:62500. DCP2 antibody can be used for detection of DCP2 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: DCP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
10108-436.pdf
Datasheet
Specifications
- Size:100 ul
- Clonality:Polyclonal
- Molecular Weight:48 kDa
- Purification:Antibody is purified by peptide affinity chromatography method.
- Form:Liquid
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Host:Rabbit
- Cat. No.:10108-436
- Reactivity:Human
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:DCP2
- Gene ID:Q8IU60
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human DCP2.
- Conjugation:Unconjugated
- Antigen Synonyms:DCP2
- Antigen Name:decapping mRNA 2
- ELISA:Yes
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store DCP2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



