Specifications
- Antibody Type:Primary
- Antigen Name:forkhead box J1
- Antigen Symbol:FOXJ1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q92949
- Antigen Synonyms:FOXJ1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:45 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store FOXJ1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human FOXJ1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10106-960
- Supplier no.:28-774
Specifications
About this item
FOXJ1 is a member of forkhead/winged-helix transcription factor family, which play crucial roles during vertebrate development. FOXJ1 may play an important role in cell fate determination during lung development and in spermatogenesis.The unique pattern of FOXJ1expression during human fetal development suggests a role for this forkhead/winged-helix factor during pulmonary and renal epithelial development. Single nucleotide polymorphisms were identified in FOXJ1 and a significant association was found with allergic rhinitis.FOXJ1 is a member of the forkhead gene family, which was originally identified in Drosophila. The forkhead family is composed of transcription factors with a conserved 100-amino acid DNA-binding motif.FOXJ1 is a member of the forkhead gene family, which was originally identified in Drosophila. The forkhead family is composed of transcription factors with a conserved 100-amino acid DNA-binding motif.
FOXJ1 antibody can be used for detection of FOXJ1 by ELISA at 1:62500. FOXJ1 antibody can be used for detection of FOXJ1 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: FOXJ1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog