Specifications
- Antibody Type:Primary
- Antigen Name:potassium voltage-gated channel, shaker-related subfamily, member 10
- Antigen Symbol:KCNA10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q16322
- Antigen Synonyms:KCNA10
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:56 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store KCNA10 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human KCNA10.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10101-102
- Supplier no.:28-252
Specifications
About this item
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Four sequence-related potassium channel genes, shaker, shaw, shab, and shal, have been identified in Drosophila, and each has been shown to have human homolog (s). KCNA10 encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. This member contains six membrane-spanning domains with a shaker-type repeat in the fourth segment. It is specifically regulated by cGMP and postulated to mediate the effects of substances that increase intracellular cGMP.
KCNA10 antibody can be used for detection of KCNA10 by ELISA at 1:312500. KCNA10 antibody can be used for detection of KCNA10 by western blot at 1.25 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: KCNA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat