- Antibody Type:Primary
- Antigen Name:ventral anterior homeobox 1
- Antigen Symbol:Vax1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:B1AVW5
- Antigen Synonyms:Vax1
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:21 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store VAX1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human VAX1.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10105-986
- Supplier no.:28-107
VAX1 is a homeo-domain containing protein from a class of homeobox transcription factors which are conserved in vertebrates. It may play an important role in the development of anterior ventral forebrain and visual system.This gene encodes a homeo-domain containing protein from a class of homeobox transcription factors which are conserved in vertebrates. Genes of this family are involved in the regulation of body development and morphogenesis. The most conserved genes, called HOX genes are found in special gene clusters. This gene belongs to the VAX subfamily and lies in the vicinity of the EMX homeobox gene family. Another member of VAX family is located on chromosome 2. The encoded protein may play an important role in the development of anterior ventral forebrain and visual system. Multiple transcript variants encoding different isoforms have been found for this gene.
VAX1 antibody can be used for detection of VAX1 by ELISA at 1:62500. VAX1 antibody can be used for detection of VAX1 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: Vax1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human