To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:cysteine-rich protein 2
- Antigen Symbol:CRIP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:P52943
- Antigen Synonyms:CRIP2
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:22 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store CRIP2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human CRIP2.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10105-908
- Supplier no.:28-067
Human ESP1/CRP2 protein has two LIM domains, and each shares 35.1% and 77 or 79% identical residues with human cysteine-rich protein (CRP) and rat CRIP, respectively. Northern blot analysis of ESP1/CRP2 in various human tissues showed distinct tissue distributions compared with CRP and CRIP, suggesting that each might serve related but specific roles in tissue organization or function. Using a panel of human-rodent somatic cell hybrids, the ESP1/CRP2 locus was assigned to chromosome 14. Fluorescence in situ hybridization, using cDNA and a genome DNA fragment of the ESP1/CRP2 as probes, confirms this assignment and relegates regional localization to band 14q32.3.
CRIP2 antibody can be used for detection of CRIP2 by ELISA at 1:62500. CRIP2 antibody can be used for detection of CRIP2 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: CRIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human