Specifications
- Antibody Type:Primary
- Antigen Name:thymocyte selection-associated high mobility group box
- Antigen Symbol:TOX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:O94900
- Antigen Synonyms:TOX
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:58 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store TOX antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human TOX.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10105-716
- Supplier no.:27-925
Specifications
About this item
Some high-mobility group (HMG) box proteins (e.g., LEF1) contain a single HMG box motif and bind DNA in a sequence-specific manner, while other members of this family (e.g., HMG1) have multiple HMG boxes and bind DNA in a sequence-independent but structure-dependent manner. All HMG box proteins are able to induce a sharp bend in DNA. TOX contains a single HMG box motif.Some high-mobility group (HMG) box proteins (e.g., LEF1; MIM 153245) contain a single HMG box motif and bind DNA in a sequence-specific manner, while other members of this family (e.g., HMG1; MIM 163905) have multiple HMG boxes and bind DNA in a sequence-independent but structure-dependent manner. All HMG box proteins are able to induce a sharp bend in DNA. TOX contains a single HMG box motif.
TOX antibody can be used for detection of TOX by ELISA at 1:312500. TOX antibody can be used for detection of TOX by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TOX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog