Anti-TEAD3 Rabbit Polyclonal Antibody
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
TEAD3 is a member of the transcriptional enhancer factor (TEF) family. The family members contain the TEA/ATTS DNA-binding domain. This member is preferentially expressed in placenta and transactivates the chorionic somatomammotropin gene enhancer.This gene encodes a member of the transcriptional enhancer factor (TEF) family. The family members contain the TEA/ATTS DNA-binding domain. This member is preferentially expressed in placenta and transactivates the chorionic somatomammotropin gene enhancer. The protein is encoded through the use of a non-AUG (ATA) translation initiation codon.
TEAD3 antibody can be used for detection of TEAD3 by ELISA at 1:312500. TEAD3 antibody can be used for detection of TEAD3 by western blot at 1.0 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TEAD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog
Documents
10105-594.pdf
Datasheet
Specifications
- Size:100 ul
- Clonality:Polyclonal
- Molecular Weight:49 kDa
- Purification:Antibody is purified by peptide affinity chromatography method.
- Form:Liquid
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Host:Rabbit
- Cat. No.:10105-594
- Reactivity:Human|Dog|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:4 °C
- Antibody Type:Primary
- Antigen Symbol:TEAD3
- Gene ID:Q99594
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human TEAD3.
- Conjugation:Unconjugated
- Antigen Synonyms:TEAD3
- Antigen Name:TEA domain family member 3
- ELISA:Yes
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store TEAD3 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



