Anti-SIRT2 Rabbit Polyclonal Antibody
Catalog # 10105-200
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:sirtuin 2
- Antigen Symbol:SIRT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q8IXJ6
- Antigen Synonyms:SIRT2
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:43 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store SIRT2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human SIRT2.
- Purification:Antibody is purified by protein A chromatography method.
- Cat. No.:10105-200
- Supplier no.:27-574
Specifications
About this item
SIRT2 is included in class I of the sirtuin family which is characterized by a sirtuin core domain. Human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity.
- SIRT2 antibody can be used for detection of SIRT2 by ELISA at 1:312500. SIRT2 antibody can be used for detection of SIRT2 by western blot at 5 μg/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SIRT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human