Specifications
- Antibody Type:Primary
- Antigen Name:T-box 19
- Antigen Symbol:TBX19
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:O60806
- Antigen Synonyms:TBX19
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:48 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store TBX19 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human TBX19.
- Purification:Antibody is supplied as total IgG.
- Cat. No.:10105-180
- Supplier no.:27-564
Specifications
About this item
TBX19 is a member of a phylogenetically conserved family of genes that share a common DNA-binding domain, the T-box. T-box genes encode transcription factors involved in the regulation of developmental processes. TBX19 is the human ortholog of mouse Tbx19/Tpit gene. Studies in mouse show that Tpit protein is present only in the two pituitary pro-opiomelanocortin (POMC)-expressing lineages, the corticotrophs and melanotrophs. Mutations in the human ortholog were found in patients with isolated deficiency of pituitary POMC-derived ACTH, suggesting an essential role for this gene in differentiation of the pituitary POMC lineage.
TBX19 antibody can be used for detection of TBX19 by ELISA at 1:62500. TBX19 antibody can be used for detection of TBX19 by western blot at 1.0-1.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is supplied as total IgG.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 200 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: TBX19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish