Specifications
- Antibody Type:Primary
- Antigen Name:general transcription factor IIi
- Antigen Symbol:GTF2I
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q75M86
- Antigen Synonyms:GTF2I
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:110 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store GTF2I antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human GTF2I.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10104-938
- Supplier no.:27-438
Specifications
About this item
GTF2I encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. The exon (s) encoding 5' UTR has not been fully defined, but this gene is known to contain at least 34 exons, and its alternative splicing generates 4 transcript variants.
GTF2I antibody can be used for detection of GTF2I by ELISA at 1:62500. GTF2I antibody can be used for detection of GTF2I by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: GTF2I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog