Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ATP6V1C1 Rabbit Polyclonal Antibody
Anti-ATP6V1C1 Rabbit Polyclonal Antibody
Catalog # 10104-576
Supplier:  Prosci
undefined
Anti-ATP6V1C1 Rabbit Polyclonal Antibody
Catalog # 10104-576
Supplier:  Prosci
Supplier Number:  27-192

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C1
  • Antigen Symbol:
    ATP6V1C1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western Blot:
    Yes
  • Size:
    100 ul
  • Form:
    Liquid
  • Gene ID:
    P21283
  • Antigen Synonyms:
    ATP6V1C1
  • Storage Buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • Molecular Weight:
    42 kDa
  • Storage Temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store ATP6V1C1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ATP6V1C1.
  • Purification:
    Antibody is purified by peptide affinity chromatography method.
  • Cat. No.:
    10104-576
  • Supplier no.:
    27-192

Specifications

About this item

ATP6V1C1 is a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D.This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of intracellular compartments of eukaryotic cells. V-ATPase dependent acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c'', and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene is one of two genes that encode the V1 domain C subunit proteins and is found ubiquitously. This C subunit is analogous but not homologous to gamma subunit of F-ATPases. Previously, this gene was designated ATP6D. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.

ATP6V1C1 antibody can be used for detection of ATP6V1C1 by ELISA at 1:312500. ATP6V1C1 antibody can be used for detection of ATP6V1C1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ATP6V1C1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat