Specifications
- Antibody Type:Primary
- Antigen Name:retinol dehydrogenase 10 (all-trans)
- Antigen Symbol:RDH10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q8IZV5
- Antigen Synonyms:RDH10
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:38 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store RDH10 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human RDH10.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10100-244
- Supplier no.:26-761
Specifications
About this item
RDH10 is a retinol dehydrogenase with a clear preference for NADP. RDH10 converts all-trans-retinol to all-trans-retinal. RDH10 has no detectable activity towards 11-cis-retinol, 9-cis-retinol and 13-cis-retinol.RDH10 generates all-trans retinal from all-trans retinol and may plan an important role in the photic visual cycle. All-trans retinal is isomerized to 11-cis retinal by the retinal G protein-coupled receptor (RGR; MIM 600342) when the retinal pigment epithelium (RPE) is illuminated.
RDH10 antibody can be used for detection of RDH10 by ELISA at 1:312500. RDH10 antibody can be used for detection of RDH10 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: RDH10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat