- Antibody Type:Primary
- Antigen Name:solute carrier family 25, member 34
- Antigen Symbol:SLC25A34
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q6PIV7
- Antigen Synonyms:SLC25A34
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:32 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store SLC25A34 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human SLC25A34.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10099-832
- Supplier no.:25-937
SLC25A34 is a multi-pass membrane protein. It belongs to the mitochondrial carrier family and contains 3 Solcar repeats. The function of the SLC25A34 protein remains unknown. SLC25A34 belongs to the SLC25 family of mitochondrial carrier proteins (Haitina et al., 2006 [PubMed 16949250]).
SLC25A34 antibody can be used for detection of SLC25A34 by ELISA at 1:312500. SLC25A34 antibody can be used for detection of SLC25A34 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SLC25A34
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse