Specifications
- Antibody Type:Primary
- Antigen Name:ATPase, H+ transporting, lysosomal V0 subunit a2
- Antigen Symbol:ATP6V0A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9Y487
- Antigen Synonyms:ATP6V0A2
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:98 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ATP6V0A2 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ATP6V0A2.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10102-586
- Supplier no.:25-762
Specifications
About this item
The multisubunit vacuolar-type proton pump (H (+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H (+)-ATPases are comprised of a peripheral V (1) domain and an integral membrane V (0) domain; ATP6V0A2 is a component of the V (0) domain.The multisubunit vacuolar-type proton pump (H (+)-ATPase or V-ATPase) is essential for acidification of diverse cellular components, including endosomes, lysosomes, clathrin-coated vesicles, secretory vesicles, and chromaffin granules, and it is found at high density in the plasma membrane of certain specialized cells. H (+)-ATPases are comprised of a peripheral V (1) domain and an integral membrane V (0) domain; ATP6V0A2 is a component of the V (0) domain (Smith et al., 2003 [PubMed 14580332]).
ATP6V0A2 antibody can be used for detection of ATP6V0A2 by ELISA at 1:312500. ATP6V0A2 antibody can be used for detection of ATP6V0A2 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ATP6V0A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat