- Antibody Type:Primary
- Antigen Name:zinc finger protein 703
- Antigen Symbol:ZNF703
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 ul
- Form:Liquid
- Gene ID:Q9H7S9
- Antigen Synonyms:ZNF703
- Storage Buffer:Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
- Molecular Weight:58 kDa
- Storage Temperature:For short periods of storage (days) store at 4 °C. For longer periods of storage, store ZNF703 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
- Shipping Temperature:4 °C
- Immunogen:Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human ZNF703.
- Purification:Antibody is purified by peptide affinity chromatography method.
- Cat. No.:10102-206
- Supplier no.:25-539
ZNF703 belongs to the Elbow/Noc family. It contains 1 C2H2-type zinc finger.ZNF703 may function as a transcriptional repressor.
ZNF703 antibody can be used for detection of ZNF703 by ELISA at 1:312500. ZNF703 antibody can be used for detection of ZNF703 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: ZNF703
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat