Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-POU1F1 Rabbit Polyclonal Antibody
Anti-POU1F1 Rabbit Polyclonal Antibody
Catalog # 10101-420
Supplier:  Prosci
undefined
Anti-POU1F1 Rabbit Polyclonal Antibody
Catalog # 10101-420
Supplier:  Prosci
Supplier Number:  25-055

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    POU class 1 homeobox 1
  • Antigen Symbol:
    POU1F1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Dog
  • Western Blot:
    Yes
  • Size:
    100 ul
  • Form:
    Liquid
  • Gene ID:
    P28069
  • Antigen Synonyms:
    POU1F1
  • Storage Buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • Molecular Weight:
    33 kDa
  • Storage Temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store POU1F1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human POU1F1.
  • Purification:
    Antibody is purified by peptide affinity chromatography method.
  • Cat. No.:
    10101-420
  • Supplier no.:
    25-055

Specifications

About this item

PIT1 is a pituitary-specific transcription factor responsible for pituitary development and hormone expression in mammals and is a member of the POU family of transcription factors that regulate mammalian development. The POU family is so named because the first 3 members identified were PIT1 and OCT1 of mammals, and Unc-86 of C. elegans. PIT1 contains 2 protein domains, termed POU-specific and POU-homeo, which are both necessary for high affinity DNA binding on genes encoding growth hormone and prolactin. PIT1 is also important for regulation of the genes encoding prolactin and thyroid-stimulating hormone beta subunit by thyrotropin-releasing hormone and cyclic AMP. This gene encodes a member of the POU family of transcription factors that regulate mammalian development. The protein regulates expression of several genes involved in pituitary development and hormone expression. Mutations in this genes result in combined pituitary hormone deficiency. Multiple transcript variants encoding different isoforms have been found for this gene.

POU1F1 antibody can be used for detection of POU1F1 by ELISA at 1:312500. POU1F1 antibody can be used for detection of POU1F1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: POU1F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Dog