Order Entry
United States
ContactUsLinkComponent
Anti-PDPK1 Rabbit Polyclonal Antibody
Anti-PDPK1 Rabbit Polyclonal Antibody
  10101-380
 :  Prosci
undefined
Anti-PDPK1 Rabbit Polyclonal Antibody
  10101-380
 :  Prosci
 :  25-028
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Name:
    pyruvate dehydrogenase kinase, isozyme 1
  • Antigen Symbol:
    PDK1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 ul
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    Q15118
  • Antigen Synonyms:
    PDK1
  • Storage Buffer:
    Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.
  • Molecular Weight:
    47 kDa
  • Storage Temperature:
    For short periods of storage (days) store at 4 °C. For longer periods of storage, store PDK1 antibody at –20 °C. As with any antibody avoid repeat freeze-thaw cycles.
  • Shipping Temperature:
    4 °C
  • Immunogen:
    Antibody produced in rabbits immunized with a synthetic peptide corresponding a region of human PDK1.
  • Purification:
    Antibody is purified by peptide affinity chromatography method.
  • Cat. No.:
    10101-380
  • Supplier no.:
    25-028

 

 

Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation.Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. Please see the Entrez Gene record to access additional publications.

PDK1 antibody can be used for detection of PDK1 by ELISA at 1:1562500. PDK1 antibody can be used for detection of PDK1 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PDK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human